Product Details

SNP ID
rs201124958
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.7:74221667 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATTGCCATGGAGTATTACAACTG[C/G]GGGCGGTTCTCGAAGCCCCCAGAAG
Phenotype
MIM: 605719 MIM: 600404
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
LAT2 PubMed Links

Gene Details

Gene
LAT2
Gene Name
linker for activation of T-cells family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014146.3 753 Missense Mutation TGC,TGG C121W NP_054865.2
NM_032463.2 753 Missense Mutation TGC,TGG C121W NP_115852.1
NM_032464.2 753 Missense Mutation TGC,TGG C121W NP_115853.2
XM_011516558.2 753 Missense Mutation TGC,TGG C121W XP_011514860.1
Gene
RFC2
Gene Name
replication factor C subunit 2
There are no transcripts associated with this gene.

View Full Product Details