Product Details

SNP ID
rs201967222
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42176111 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGATGATGCCCACCAAAGTCATG[C/T]GGCCATCGTTCAGACACACCAGGGG
Phenotype
MIM: 610469 MIM: 173370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AP3M2 PubMed Links

Gene Details

Gene
AP3M2
Gene Name
adaptor related protein complex 3 mu 2 subunit
There are no transcripts associated with this gene.

Gene
PLAT
Gene Name
plasminogen activator, tissue type
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000930.4 1690 Missense Mutation CAC,CGC H524R NP_000921.1
NM_001319189.1 1690 Missense Mutation CAC,CGC H435R NP_001306118.1
NM_033011.3 1690 Missense Mutation CAC,CGC H478R NP_127509.1

View Full Product Details