Product Details
- SNP ID
-
rs201949500
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:102829909 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ACCATCATTCATATAATACATGAAG[A/G]CTGGTTCATCACTTCCGGTTTTTTC
- Phenotype
-
MIM: 607909
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AZIN1
PubMed Links
Gene Details
- Gene
- AZIN1
- Gene Name
- antizyme inhibitor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001301668.1 |
1559 |
Missense Mutation |
GCC,GTC |
A311V |
NP_001288597.1 |
NM_015878.5 |
1559 |
Missense Mutation |
GCC,GTC |
A311V |
NP_056962.2 |
NM_148174.3 |
1559 |
Missense Mutation |
GCC,GTC |
A311V |
NP_680479.1 |
XM_005250969.1 |
1559 |
Missense Mutation |
GCC,GTC |
A310V |
XP_005251026.1 |
XM_011517126.1 |
1559 |
Missense Mutation |
GCC,GTC |
A311V |
XP_011515428.1 |
XM_011517127.1 |
1559 |
Missense Mutation |
GCC,GTC |
A311V |
XP_011515429.1 |
XM_011517128.2 |
1559 |
Missense Mutation |
GCC,GTC |
A144V |
XP_011515430.1 |
XM_017013563.1 |
1559 |
Missense Mutation |
GCC,GTC |
A310V |
XP_016869052.1 |
XM_017013564.1 |
1559 |
Missense Mutation |
GCC,GTC |
A310V |
XP_016869053.1 |
XM_017013565.1 |
1559 |
Missense Mutation |
GCC,GTC |
A144V |
XP_016869054.1 |
XM_017013566.1 |
1559 |
Missense Mutation |
GCC,GTC |
A143V |
XP_016869055.1 |
- Gene
- LOC107986873
- Gene Name
- forkhead box protein L2-like
View Full Product Details