Product Details

SNP ID
rs202036744
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:43103684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTCGACCACTTCTTCATCGCTCCT[C/T]GACAATCCACTGTGGACCCCACACA
Phenotype
MIM: 615247
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
POMK PubMed Links

Gene Details

Gene
POMK
Gene Name
protein-O-mannose kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001277971.1 307 Nonsense Mutation CGA,TGA R46* NP_001264900.1
NM_032237.4 307 Nonsense Mutation CGA,TGA R46* NP_115613.1

View Full Product Details