Product Details

SNP ID
rs201468098
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:98428037 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCTAACGTCGAGGACGGGGAGATC[C/T]GCATCAATGTGGGCGGCTTCAAGAG
Phenotype
MIM: 602906 MIM: 605030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KCNS2 PubMed Links

Gene Details

Gene
KCNS2
Gene Name
potassium voltage-gated channel modifier subfamily S member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020697.3 408 Missense Mutation CGC,TGC R20C NP_065748.1
Gene
STK3
Gene Name
serine/threonine kinase 3
There are no transcripts associated with this gene.

View Full Product Details