Product Details

SNP ID
rs202165364
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:51320014 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACAAATCTCCACCACACAGGTGAC[A/C]TGGCCACTCTGAAAGGCAGCATGCA
Phenotype
MIM: 615904
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PXDNL PubMed Links

Gene Details

Gene
PXDNL
Gene Name
peroxidasin like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_144651.4 5668 Missense Mutation NP_653252.3
XM_005251168.3 5668 Missense Mutation XP_005251225.1
XM_006716420.3 5668 Missense Mutation XP_006716483.1
XM_011517456.2 5668 Missense Mutation XP_011515758.1
XM_011517457.2 5668 Missense Mutation XP_011515759.1
XM_011517458.2 5668 Missense Mutation XP_011515760.1
XM_011517459.2 5668 Intron XP_011515761.1
XM_017013041.1 5668 Missense Mutation XP_016868530.1

View Full Product Details