Product Details

SNP ID
rs200775391
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:94250450 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTTTTCTGGTAGGCCAGCCGCC[A/G]TTCATTCTTCTCCTTGCTGTCCCGC
Phenotype
MIM: 600164
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GEM PubMed Links

Gene Details

Gene
GEM
Gene Name
GTP binding protein overexpressed in skeletal muscle
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005261.3 1194 Missense Mutation CGG,TGG R251W NP_005252.1
NM_181702.2 1194 Missense Mutation CGG,TGG R251W NP_859053.1
XM_017013315.1 1194 UTR 3 XP_016868804.1

View Full Product Details