Product Details

SNP ID
rs200001487
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:109340533 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTAGAACACGTTACTGTTGATGAC[C/T]TGGTGGCTGAAATCACTCCAAAAGG
Phenotype
MIM: 606109
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ENY2 PubMed Links

Gene Details

Gene
ENY2
Gene Name
ENY2, transcription and export complex 2 subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193557.1 273 Silent Mutation CTG,TTG L62L NP_001180486.1
NM_020189.5 273 Silent Mutation CTG,TTG L67L NP_064574.1
Gene
NUDCD1
Gene Name
NudC domain containing 1
There are no transcripts associated with this gene.

View Full Product Details