Product Details

SNP ID
rs200260363
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:123645914 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCAGCCGCCGCCCGCCCGTCACG[C/T]AGAGTTTGTTTCCCATCACTGTGGC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KLHL38 PubMed Links

Gene Details

Gene
KLHL38
Gene Name
kelch like family member 38
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081675.2 1816 Missense Mutation TAC,TGC Y524C NP_001075144.2
XM_005250901.4 1816 Missense Mutation TAC,TGC Y524C XP_005250958.1

View Full Product Details