Product Details

SNP ID
rs200134502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:143991579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAACCCCTATGGACAGCCACAGG[A/T]CTTCCCAGGACAAGACCCTGACTGT
Phenotype
MIM: 138251 MIM: 609564
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
GRINA PubMed Links

Gene Details

Gene
GRINA
Gene Name
glutamate ionotropic receptor NMDA type subunit associated protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000837.1 522 Missense Mutation GAC,GTC D119V NP_000828.1
NM_001009184.1 522 Missense Mutation GAC,GTC D119V NP_001009184.1
Gene
PARP10
Gene Name
poly(ADP-ribose) polymerase family member 10
There are no transcripts associated with this gene.

View Full Product Details