Product Details

SNP ID
rs200585226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:96145448 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAAGAGCCGCAGCTCCGCGCCCAC[A/C]AGCTCTTCTTTGTCTGAGAGCATGG
Phenotype
MIM: 601147
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GDF6 PubMed Links

Gene Details

Gene
GDF6
Gene Name
growth differentiation factor 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001557.2 583 Silent Mutation CTG,CTT L161L NP_001001557.1

View Full Product Details