Product Details

SNP ID
rs200310088
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:23137942 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTTTTCGGAGCCCACCAGTTGGTC[C/T]TGAATTGTTTCCTTTGCATGTCCTT
Phenotype
MIM: 603614
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TNFRSF10D PubMed Links

Gene Details

Gene
TNFRSF10D
Gene Name
TNF receptor superfamily member 10d
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003840.4 1184 Silent Mutation CAA,CAG Q363Q NP_003831.2

View Full Product Details