Product Details

SNP ID
rs200899389
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137244194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGGGGATGAACCCCATGTAGAAG[C/G]GGATCAGGCCTTGGCTGCTGTAGAT
Phenotype
MIM: 609826 MIM: 602660
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
C9orf173-AS1 PubMed Links

Gene Details

Gene
C9orf173-AS1
Gene Name
C9orf173 antisense RNA 1
There are no transcripts associated with this gene.

Gene
FAM166A
Gene Name
family with sequence similarity 166 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001710.2 1759 Missense Mutation CCC,CGC P281R NP_001001710.1
XM_006717108.2 1759 Missense Mutation CCC,CGC P265R XP_006717171.2
XM_011518694.1 1759 Missense Mutation CCC,CGC P316R XP_011516996.1
XM_011518695.1 1759 Missense Mutation CCC,CGC P278R XP_011516997.1
XM_011518696.2 1759 Intron XP_011516998.1
XM_011518697.1 1759 Intron XP_011516999.1
XM_017014718.1 1759 Missense Mutation CCC,CGC P194R XP_016870207.1
Gene
SLC34A3
Gene Name
solute carrier family 34 member 3
There are no transcripts associated with this gene.

Gene
STPG3
Gene Name
sperm-tail PG-rich repeat containing 3
There are no transcripts associated with this gene.

Gene
TUBB4B
Gene Name
tubulin beta 4B class IVb
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006088.5 1759 Intron NP_006079.1

View Full Product Details