Product Details

SNP ID
rs200511236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:129009820 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCATGTGGCGGTAGCACTGTGCGT[A/C]GTAGGTTGTCTGAGACTTGACGAAC
Phenotype
MIM: 615587 MIM: 609288
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
NUP188 PubMed Links

Gene Details

Gene
NUP188
Gene Name
nucleoporin 188
There are no transcripts associated with this gene.

Gene
SH3GLB2
Gene Name
SH3 domain containing GRB2 like endophilin B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001287045.1 1045 Missense Mutation GAC,TAC D268Y NP_001273974.1
NM_001287046.1 1045 Missense Mutation GAC,TAC D264Y NP_001273975.1
NM_020145.3 1045 Missense Mutation GAC,TAC D264Y NP_064530.1
XM_005252098.2 1045 Missense Mutation GAC,TAC D264Y XP_005252155.1
XM_005252100.2 1045 Missense Mutation GAC,TAC D243Y XP_005252157.1
XM_005252101.3 1045 UTR 5 XP_005252158.1
XM_006717188.2 1045 Missense Mutation GAC,TAC D292Y XP_006717251.1
XM_006717189.2 1045 Missense Mutation GAC,TAC D288Y XP_006717252.1
XM_006717190.1 1045 Missense Mutation GAC,TAC D292Y XP_006717253.1
XM_006717191.1 1045 Missense Mutation GAC,TAC D268Y XP_006717254.1

View Full Product Details