Product Details

SNP ID
rs201209437
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109381684 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCACCATCATGGCGCGCTGGTCTC[A/G]CATTTTTCGGACAATATCCAGTGGG
Phenotype
MIM: 176877
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PTPN3 PubMed Links

Gene Details

Gene
PTPN3
Gene Name
protein tyrosine phosphatase, non-receptor type 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145368.1 3484 Silent Mutation NP_001138840.1
NM_001145369.1 3484 Silent Mutation NP_001138841.1
NM_001145370.1 3484 Silent Mutation NP_001138842.1
NM_001145371.1 3484 Silent Mutation NP_001138843.1
NM_001145372.1 3484 Silent Mutation NP_001138844.1
NM_002829.3 3484 Silent Mutation NP_002820.3
XM_006717197.3 3484 Nonsense Mutation XP_006717260.1
XM_006717199.3 3484 Nonsense Mutation XP_006717262.1
XM_006717201.3 3484 Nonsense Mutation XP_006717264.1
XM_006717202.3 3484 Nonsense Mutation XP_006717265.1
XM_006717203.3 3484 Nonsense Mutation XP_006717266.1
XM_006717204.3 3484 Nonsense Mutation XP_006717267.1
XM_011518888.2 3484 Nonsense Mutation XP_011517190.1
XM_011518889.2 3484 Nonsense Mutation XP_011517191.1
XM_017014955.1 3484 Nonsense Mutation XP_016870444.1
XM_017014956.1 3484 Nonsense Mutation XP_016870445.1
XM_017014957.1 3484 Nonsense Mutation XP_016870446.1

View Full Product Details