Product Details

SNP ID
rs200251028
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:81991514 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCAACAATCAAAGGCATTGACCA[C/T]TCACACCTTGCATCTTCAGAATTCA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SPATA31D1 PubMed Links

Gene Details

Gene
SPATA31D1
Gene Name
SPATA31 subfamily D member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001670.2 1069 Silent Mutation CAC,CAT H348H NP_001001670.1
XM_017014710.1 1069 Silent Mutation CAC,CAT H311H XP_016870199.1

View Full Product Details