Product Details
- SNP ID
-
rs200399023
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:136763467 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATGCGCACGTCCAGGTAGCCCAAG[A/G]CTGCGGAGAGGCAGGCGGCCTTTTC
- Phenotype
-
MIM: 612902
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LCN15
PubMed Links
Gene Details
- Gene
- LCN15
- Gene Name
- lipocalin 15
- Gene
- LCN8
- Gene Name
- lipocalin 8
There are no transcripts associated with this gene.
View Full Product Details