Product Details

SNP ID
rs200892914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:131579569 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTAATTTTCAGAGACAGTTCCCA[A/T]AGGGCCTCCTCAGCCAGGTGGTCAC
Phenotype
MIM: 600303
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
RAPGEF1 PubMed Links

Gene Details

Gene
RAPGEF1
Gene Name
Rap guanine nucleotide exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304275.1 3375 Silent Mutation CTA,CTT L1071L NP_001291204.1
NM_005312.3 3375 Silent Mutation CTA,CTT L1054L NP_005303.2
NM_198679.1 3375 Silent Mutation CTA,CTT L1072L NP_941372.1
XM_005272186.4 3375 Silent Mutation CTA,CTT L1241L XP_005272243.1
XM_005272191.3 3375 Silent Mutation CTA,CTT L1203L XP_005272248.1
XM_006717067.3 3375 Silent Mutation CTA,CTT L1247L XP_006717130.1
XM_006717072.3 3375 Silent Mutation CTA,CTT L1209L XP_006717135.1
XM_006717074.3 3375 Silent Mutation CTA,CTT L1077L XP_006717137.1
XM_011518569.2 3375 Silent Mutation CTA,CTT L1255L XP_011516871.1
XM_011518570.2 3375 Silent Mutation CTA,CTT L1254L XP_011516872.1
XM_011518571.2 3375 Silent Mutation CTA,CTT L1250L XP_011516873.1
XM_011518572.2 3375 Silent Mutation CTA,CTT L1249L XP_011516874.1
XM_011518573.2 3375 Silent Mutation CTA,CTT L1232L XP_011516875.1
XM_011518574.2 3375 Silent Mutation CTA,CTT L1224L XP_011516876.1
XM_011518575.2 3375 Silent Mutation CTA,CTT L1223L XP_011516877.1
XM_011518576.2 3375 Silent Mutation CTA,CTT L1138L XP_011516878.1
XM_011518577.2 3375 Silent Mutation CTA,CTT L1137L XP_011516879.1
XM_011518578.2 3375 Silent Mutation CTA,CTT L1116L XP_011516880.1
XM_011518579.2 3375 Silent Mutation CTA,CTT L1085L XP_011516881.1
XM_011518580.2 3375 Intron XP_011516882.1
XM_011518581.2 3375 Intron XP_011516883.1
XM_011518582.2 3375 Intron XP_011516884.1
XM_017014633.1 3375 Silent Mutation CTA,CTT L1224L XP_016870122.1
XM_017014634.1 3375 Silent Mutation CTA,CTT L1218L XP_016870123.1
XM_017014635.1 3375 Silent Mutation CTA,CTT L1218L XP_016870124.1
XM_017014636.1 3375 Silent Mutation CTA,CTT L1210L XP_016870125.1
XM_017014637.1 3375 Silent Mutation CTA,CTT L1202L XP_016870126.1
XM_017014638.1 3375 Silent Mutation CTA,CTT L1171L XP_016870127.1
XM_017014639.1 3375 Silent Mutation CTA,CTT L1169L XP_016870128.1
XM_017014640.1 3375 Intron XP_016870129.1
XM_017014641.1 3375 Intron XP_016870130.1

View Full Product Details