Product Details

SNP ID
rs201119765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33442475 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGACGGGGTTGTTGTAGGGGTCA[A/G]CAATGGCCAGCACACACACGATAAG
Phenotype
MIM: 600170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AQP3 PubMed Links

Gene Details

Gene
AQP3
Gene Name
aquaporin 3 (Gill blood group)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001318144.1 639 Intron NP_001305073.1
NM_004925.4 639 Missense Mutation GCT,GTT A179V NP_004916.1

View Full Product Details