Product Details

SNP ID
rs201243209
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128260108 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGCTCGCTCAGCTTCTCCTGCA[A/G]CTCGCCCAGCTTCTTTCCCAGCTCC
Phenotype
MIM: 602377 MIM: 602580
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DNM1 PubMed Links

Gene Details

Gene
DNM1
Gene Name
dynamin 1
There are no transcripts associated with this gene.

Gene
GOLGA2
Gene Name
golgin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004486.4 Intron NP_004477.3
XM_005251930.1 Intron XP_005251987.1
XM_005251931.1 Intron XP_005251988.1
XM_005251932.1 Intron XP_005251989.1

View Full Product Details