Product Details

SNP ID
rs200403260
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1345328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCACCGTTTTAGATCCAAACCCAC[C/T]AATCACGAACCTAAGGATGAAAGCA
Phenotype
MIM: 308385 MIM: 430000
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL3RA PubMed Links

Gene Details

Gene
IL3RA
Gene Name
interleukin 3 receptor subunit alpha
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267713.1 464 Intron NP_001254642.1
NM_002183.3 464 Missense Mutation CCA,CTA P26L NP_002174.1
XM_005274431.4 464 Missense Mutation CCA,CTA P26L XP_005274488.1
XM_005274432.1 464 Missense Mutation CCA,CTA P26L XP_005274489.1
XM_017029491.1 464 Missense Mutation CCA,CTA P26L XP_016884980.1

View Full Product Details