Product Details
- SNP ID
-
rs199829241
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:137879099 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGGCCACCATCATCGCCAACTGCAT[C/T]GTGCTGGCCCTGGAGCAGCACCTCC
- Phenotype
-
MIM: 601012
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CACNA1B
PubMed Links
Gene Details
- Gene
- CACNA1B
- Gene Name
- calcium voltage-gated channel subunit alpha1 B
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000718.3 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
NP_000709.1 |
NM_001243812.1 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
NP_001230741.1 |
XM_011518991.2 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_011517293.1 |
XM_011518992.2 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_011517294.1 |
XM_011518993.2 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_011517295.1 |
XM_011518994.2 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_011517296.1 |
XM_017015115.1 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_016870604.1 |
XM_017015116.1 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_016870605.1 |
XM_017015117.1 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_016870606.1 |
XM_017015118.1 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_016870607.1 |
XM_017015119.1 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_016870608.1 |
XM_017015120.1 |
438 |
Silent Mutation |
ATC,ATT |
I110I |
XP_016870609.1 |
- Gene
- LOC100133077
- Gene Name
- uncharacterized LOC100133077
There are no transcripts associated with this gene.
View Full Product Details