Product Details

SNP ID
rs199731037
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:109137597 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGACAACACGCTCTGAAGCCGGC[A/G]GTGAGTCTATATCATGTCGAGTGAT
Phenotype
MIM: 604574
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FRRS1L PubMed Links

Gene Details

Gene
FRRS1L
Gene Name
ferric chelate reductase 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011518453.2 1277 Missense Mutation CCG,CTG P182L XP_011516755.1
XM_011518454.2 1277 Missense Mutation CCG,CTG P182L XP_011516756.1

View Full Product Details