Product Details

SNP ID
rs201552425
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:96450339 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGGCGCCGCGATGCAGGAGAGTTT[C/T]GGCTGCGTGGTGGCCAACCGCTTCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HABP4 PubMed Links

Gene Details

Gene
HABP4
Gene Name
hyaluronan binding protein 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014282.3 185 Silent Mutation TTC,TTT F20F NP_055097.2
XM_005251812.3 185 Intron XP_005251869.1
XM_017014484.1 185 Intron XP_016869973.1

View Full Product Details