Product Details

SNP ID
rs201536301
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128149580 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCTGTTGGGGGCTCTGCATGCC[C/T]AGGCCCAGGACTCCACCTCAGACCT
Phenotype
MIM: 600181
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C9orf16 PubMed Links

Gene Details

Gene
C9orf16
Gene Name
chromosome 9 open reading frame 16
There are no transcripts associated with this gene.

Gene
LCN2
Gene Name
lipocalin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005564.4 151 Nonsense Mutation CAG,TAG Q19* NP_005555.2

View Full Product Details