Product Details
- SNP ID
-
rs201950652
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:22447184 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCCGGCGTTGCCGGTACCATCGG[C/G]GATGCAGGTTCCCCCAGCGTTCCTG
- Phenotype
-
MIM: 614803
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
DMRTA1
PubMed Links
Gene Details
- Gene
- DMRTA1
- Gene Name
- DMRT like family A1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022160.2 |
344 |
Missense Mutation |
GCG,GGG |
A40G |
NP_071443.2 |
View Full Product Details