Product Details
- SNP ID
-
rs201973871
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:77020729 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCTGTGCCACTCGGCAAGCCAGAGC[C/G]TGCCTGCCATGCCGGTGCCCATCAA
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
FOXB2
PubMed Links
Gene Details
- Gene
- FOXB2
- Gene Name
- forkhead box B2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001013735.1 |
1075 |
Missense Mutation |
CTG,GTG |
L359V |
NP_001013757.1 |
View Full Product Details