Product Details

SNP ID
rs200045736
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:55486818 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTGGTAAGACTAGTCTTTCTCTA[C/G]ACCCTGTTTGTGATAGAACAGTAAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
USP51 PubMed Links

Gene Details

Gene
USP51
Gene Name
ubiquitin specific peptidase 51
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_201286.3 2201 Missense Mutation CTA,GTA L708V NP_958443.1
XM_017029299.1 2201 Missense Mutation CTA,GTA L708V XP_016884788.1
XM_017029300.1 2201 Missense Mutation CTA,GTA L708V XP_016884789.1
XM_017029301.1 2201 Missense Mutation CTA,GTA L708V XP_016884790.1

View Full Product Details