Product Details

SNP ID
rs199503317
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:106942097 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAATCTGGAATTCAGCTTGGCGA[C/T]AACCCATGTGTTTCTTGCTCCAGTA
Phenotype
MIM: 300970
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MORC4 PubMed Links

Gene Details

Gene
MORC4
Gene Name
MORC family CW-type zinc finger 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085354.2 2776 Missense Mutation TAT,TGT Y834C NP_001078823.1
NM_024657.4 2776 Missense Mutation TAT,TGT Y834C NP_078933.3
XM_005262190.3 2776 Missense Mutation TAT,TGT Y742C XP_005262247.1
XM_006724691.2 2776 Missense Mutation TAT,TGT Y712C XP_006724754.1
XM_011531027.2 2776 Missense Mutation TAT,TGT Y657C XP_011529329.1
XM_017029844.1 2776 Missense Mutation TAT,TGT Y742C XP_016885333.1

View Full Product Details