Product Details

SNP ID
rs200211063
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30560224 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTGACTTGTGTTTCATCCACAGA[A/G]GAATAGGAAAGGGTAGCAACAGAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXorf21 PubMed Links

Gene Details

Gene
CXorf21
Gene Name
chromosome X open reading frame 21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025159.2 527 Silent Mutation TCC,TCT S44S NP_079435.1

View Full Product Details