Product Details

SNP ID
rs199953225
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:75274565 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCTAGAGGACTGCGAACTCTCCC[A/G]GCAGATCGGGGCGCAGCTTAAGCTG
Phenotype
MIM: 300656
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
UPRT PubMed Links

Gene Details

Gene
UPRT
Gene Name
uracil phosphoribosyltransferase homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001307944.1 507 Missense Mutation CAG,CGG Q104R NP_001294873.1
NM_145052.3 507 Missense Mutation CAG,CGG Q104R NP_659489.1
XM_011530867.2 507 Missense Mutation CAG,CGG Q104R XP_011529169.1

View Full Product Details