Product Details

SNP ID
rs199775125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:47586579 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGTGGCACTCATTGCTTGTGGA[C/T]GGACCAGCTCCTCCAAGGCTCTGAA
Phenotype
MIM: 313440 MIM: 305370
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MIR4769 PubMed Links

Gene Details

Gene
MIR4769
Gene Name
microRNA 4769
There are no transcripts associated with this gene.

Gene
SYN1
Gene Name
synapsin I
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006950.3 704 Intron NP_008881.2
NM_133499.2 704 Intron NP_598006.1
Gene
TIMP1
Gene Name
TIMP metallopeptidase inhibitor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003254.2 704 Missense Mutation ACG,ATG T171M NP_003245.1
XM_017029766.1 704 Intron XP_016885255.1

View Full Product Details