Product Details
- SNP ID
-
rs199644815
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:67545218 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGACCTACCGAGGAGCTTTCCA[A/G]AATCTGTTCCAGAGCGTGCGCGAAG
- Phenotype
-
MIM: 313700
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
AR
PubMed Links
Gene Details
- Gene
- AR
- Gene Name
- androgen receptor
View Full Product Details