Product Details

SNP ID
rs202132872
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:56563894 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGCCATGGCTGAGAATGGCGAGAG[C/G]AGCGGCCCCCCGCGCCCCTCCCGCG
Phenotype
MIM: 300264
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
UBQLN2 PubMed Links

Gene Details

Gene
UBQLN2
Gene Name
ubiquilin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_013444.3 302 Missense Mutation AGC,AGG S7R NP_038472.2

View Full Product Details