Product Details

SNP ID
rs202213679
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54449194 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCGTTTGGTGATAGAATTGAAGGG[C/T]TCCTGGCAGCGCATGCACATGGTGA
Phenotype
MIM: 300546 MIM: 300945
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
FGD1 PubMed Links

Gene Details

Gene
FGD1
Gene Name
FYVE, RhoGEF and PH domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004463.2 2957 Silent Mutation GAA,GAG E741E NP_004454.2
Gene
TSR2
Gene Name
TSR2, ribosome maturation factor
There are no transcripts associated with this gene.

View Full Product Details