Product Details

SNP ID
rs202200283
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:153057642 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTGAGATGCTACAGATGTGGCAGG[G/T]GCCCGAGGGGGAAAAGAGGCGGAGG
Phenotype
MIM: 300675
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PNMA3 PubMed Links

Gene Details

Gene
PNMA3
Gene Name
paraneoplastic Ma antigen 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282535.1 923 Missense Mutation GGG,GTG G196V NP_001269464.1
NM_013364.5 923 Missense Mutation GGG,GTG G196V NP_037496.4

View Full Product Details