Product Details
- SNP ID
-
rs201550677
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:73447464 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGATCCTCACTGGCCGTCTCTGGGA[G/T]TCTGGGGCTCACCCTACAGTCCCCC
- Phenotype
-
MIM: 300025
- Polymorphism
- G/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CDX4
PubMed Links
Gene Details
- Gene
- CDX4
- Gene Name
- caudal type homeobox 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_005193.1 |
211 |
Missense Mutation |
GTC,TTC |
V71F |
NP_005184.1 |
View Full Product Details