Product Details

SNP ID
rs201579827
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:71096685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGAGCCGCCCGAGGTGGAGCCA[C/G]ATCTGGGGGAAAAGGTACACACGGA
Phenotype
MIM: 300033
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
CXorf65 PubMed Links

Gene Details

Gene
CXorf65
Gene Name
chromosome X open reading frame 65
There are no transcripts associated with this gene.

Gene
FOXO4
Gene Name
forkhead box O4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170931.1 537 Missense Mutation CAT,GAT H53D NP_001164402.1
NM_005938.3 537 Missense Mutation CAT,GAT H53D NP_005929.2
XM_011530957.2 537 Intron XP_011529259.1

View Full Product Details