Product Details

SNP ID
rs200509545
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:30250854 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGAATGCTGATGCACTTCATTCTA[C/T]GTAAGTATAAAATGAGAGAGCCCAT
Phenotype
MIM: 300097 MIM: 300153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAGEB1 PubMed Links

Gene Details

Gene
MAGEB1
Gene Name
MAGE family member B1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002363.4 524 Missense Mutation CGT,TGT R121C NP_002354.2
NM_177404.2 524 Missense Mutation CGT,TGT R121C NP_796379.1
NM_177415.2 524 Missense Mutation CGT,TGT R121C NP_803134.1
Gene
MAGEB4
Gene Name
MAGE family member B4
There are no transcripts associated with this gene.

View Full Product Details