Product Details

SNP ID
rs201501755
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:54198344 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGATTCATTTAGGACCAGGAGGGA[C/T]TGTGGCAGGATTCTGCACTGATGAA
Phenotype
MIM: 300358
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
WNK3 PubMed Links

Gene Details

Gene
WNK3
Gene Name
WNK lysine deficient protein kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002838.3 5822 Missense Mutation ATC,GTC I1738V NP_001002838.1
NM_020922.4 5822 Missense Mutation ATC,GTC I1795V NP_065973.2
XM_006724593.3 5822 Missense Mutation ATC,GTC I1738V XP_006724656.1
XM_011530802.2 5822 Missense Mutation ATC,GTC I1748V XP_011529104.1
XM_017029741.1 5822 Missense Mutation ATC,GTC I1795V XP_016885230.1
XM_017029742.1 5822 Missense Mutation ATC,GTC I1795V XP_016885231.1
XM_017029743.1 5822 Missense Mutation ATC,GTC I1795V XP_016885232.1
XM_017029744.1 5822 Missense Mutation ATC,GTC I1795V XP_016885233.1
XM_017029745.1 5822 Missense Mutation ATC,GTC I1785V XP_016885234.1

View Full Product Details