Product Details

SNP ID
rs201111874
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:46853500 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTTGATCCAAAAGACTACATGTTC[A/G]GTGGACTGAAGGATGAAACAGTAGG
Phenotype
MIM: 300757
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
RP2 PubMed Links

Gene Details

Gene
RP2
Gene Name
retinitis pigmentosa 2 (X-linked recessive)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006915.2 316 Missense Mutation AGT,GGT S43G NP_008846.2

View Full Product Details