Product Details

SNP ID
rs201849067
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:93672385 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAATAGTCAGGAATGCCTTTAGGG[A/T]CTTCTCTTTTAGGCTTTCCCTTCCT
Phenotype
MIM: 300117
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM133A PubMed Links

Gene Details

Gene
FAM133A
Gene Name
family with sequence similarity 133 member A
There are no transcripts associated with this gene.

Gene
NAP1L3
Gene Name
nucleosome assembly protein 1 like 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004538.5 1299 Missense Mutation GAC,GTC D307V NP_004529.2

View Full Product Details