Product Details

SNP ID
rs201823249
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:110003966 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCTGCGGCACATCCTGAGGCAGAC[A/G]AAGGAGGACGGTAGGGGTAGCCCTG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TMEM164 PubMed Links

Gene Details

Gene
TMEM164
Gene Name
transmembrane protein 164
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017698.2 826 Intron NP_060168.2
NM_032227.3 826 Silent Mutation ACA,ACG T64T NP_115603.2
XM_005262205.4 826 Silent Mutation ACA,ACG T64T XP_005262262.1
XM_005262206.3 826 Silent Mutation ACA,ACG T64T XP_005262263.1
XM_005262208.4 826 Intron XP_005262265.1
XM_011531051.2 826 UTR 5 XP_011529353.1
XM_011531054.1 826 Intron XP_011529356.1
XM_011531055.2 826 Intron XP_011529357.1
XM_017029894.1 826 Intron XP_016885383.1
XM_017029895.1 826 Silent Mutation ACA,ACG T64T XP_016885384.1
XM_017029896.1 826 UTR 5 XP_016885385.1
XM_017029897.1 826 Intron XP_016885386.1
XM_017029898.1 826 Silent Mutation ACA,ACG T64T XP_016885387.1
XM_017029899.1 826 UTR 5 XP_016885388.1
XM_017029900.1 826 UTR 5 XP_016885389.1

View Full Product Details