Product Details

SNP ID
rs200785684
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:137030462 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCAGGGATAGCACATAGGAGAAAA[C/T]GATGATGAAGATCACTTTAGCAGCT
Phenotype
MIM: 300393
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR101 PubMed Links

Gene Details

Gene
GPR101
Gene Name
G protein-coupled receptor 101
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_054021.1 1213 Missense Mutation ATT,GTT I405V NP_473362.1

View Full Product Details