Product Details

SNP ID
rs200811997
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:43769410 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGTAAATCCTGTCCACTGGCTGG[C/G]GTAGAACCCTTGGAACAAAACCAAA
Phenotype
MIM: 309860
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAOB PubMed Links

Gene Details

Gene
MAOB
Gene Name
monoamine oxidase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000898.4 1380 Missense Mutation CCC,CGC P415R NP_000889.3
XM_005272608.3 1380 Missense Mutation CCC,CGC P399R XP_005272665.1
XM_017029523.1 1380 Missense Mutation CCC,CGC P399R XP_016885012.1
XM_017029524.1 1380 Missense Mutation CCC,CGC P399R XP_016885013.1

View Full Product Details