Product Details

SNP ID
rs2065663
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:149760331 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GTACATTGTTCAGTTGCTATAAAAA[A/G]AGACCAAACCACAATACCTTAAACA
Phenotype
MIM: 176851
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCMT1 PubMed Links

Gene Details

Gene
PCMT1
Gene Name
protein-L-isoaspartate (D-aspartate) O-methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252049.1 Intron NP_001238978.1
NM_001252050.1 Intron NP_001238979.1
NM_001252051.1 Intron NP_001238980.1
NM_001252052.1 Intron NP_001238981.1
NM_001252053.1 Intron NP_001238982.1
NM_005389.2 Intron NP_005380.2
XM_011535868.2 Intron XP_011534170.1

View Full Product Details