Product Details

SNP ID
rs10507174
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:104128302 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCTTTTTAGACTTTCTCATTGAACC[A/G]TAACACAAGATTGTAAATTTTCCTA
Phenotype
MIM: 189904
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NFYB PubMed Links

Gene Details

Gene
NFYB
Gene Name
nuclear transcription factor Y subunit beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006166.3 Intron NP_006157.1
XM_005268908.4 Intron XP_005268965.1
XM_005268909.4 Intron XP_005268966.1
XM_005268910.4 Intron XP_005268967.1

View Full Product Details