Product Details

SNP ID
rs2028891
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:165211148 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CAGAAAATAACTACTCTTTTTTTAA[G/T]TTTACAAAGGTAAATTGACAAATGC
Phenotype
MIM: 182390 MIM: 182391
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
SCN2A PubMed Links

Gene Details

Gene
SCN2A
Gene Name
sodium voltage-gated channel alpha subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040142.1 Intron NP_001035232.1
NM_001040143.1 Intron NP_001035233.1
NM_021007.2 Intron NP_066287.2
XM_005246753.3 Intron XP_005246810.1
XM_005246755.4 Intron XP_005246812.1
XM_017004655.1 Intron XP_016860144.1
XM_017004656.1 Intron XP_016860145.1
XM_017004657.1 Intron XP_016860146.1
XM_017004658.1 Intron XP_016860147.1
XM_017004659.1 Intron XP_016860148.1
Gene
SCN3A
Gene Name
sodium voltage-gated channel alpha subunit 3
There are no transcripts associated with this gene.

View Full Product Details