Product Details
- SNP ID
-
rs6130953
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.20:45867349 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCACACAAAAGAACAGTTGGAGATT[A/G]GTCAGATGCTCCAGTCACTGTAGTG
- Phenotype
-
MIM: 608123
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ACOT8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74274802] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ACOT8
- Gene Name
- acyl-CoA thioesterase 8
There are no transcripts associated with this gene.
- Gene
- ZSWIM3
- Gene Name
- zinc finger SWIM-type containing 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_080752.3 |
|
Intron |
|
|
NP_542790.2 |
View Full Product Details