Product Details

SNP ID
rs9526823
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52026917 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATCGCCTGGGGAATATGTACAGAG[C/T]GAAAAGAGGCTCTCAGTACAAAATG
Phenotype
MIM: 613666 MIM: 608969
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALG11 PubMed Links

Gene Details

Gene
ALG11
Gene Name
ALG11, alpha-1,2-mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004127.2 Intron NP_001004127.2
Gene
LOC107984562
Gene Name
serine/threonine-protein kinase Nek5-like
There are no transcripts associated with this gene.

Gene
UTP14C
Gene Name
UTP14, small subunit processome component homolog C (S. cerevisiae)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_021645.5 Intron NP_067677.4

View Full Product Details